missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Heptafluorobutyrylimidazole, ≥98.5% (GC); 98.5-101.0 wt. % (NT), MilliporeSigma™ Supelco™
$127.03 - $888.70
Chemical Identifiers
| MDL Number | MFCD00014503 |
|---|---|
| Synonym | 1-(Perfluorobutyryl)imidazole; NSC 151966 |
Chemical Identifiers
| MFCD00014503 | |
| 1-(Perfluorobutyryl)imidazole; NSC 151966 |
Specifications
| 32477-35-3 | |
| 161°C | |
| C7H3F7N2 O | |
| 1 mL | |
| 264.10 | |
| For GC derivatization |
| 1.490 g/mL (at 25°C) | |
| 77°C - Closed Cup | |
| MFCD00014503 | |
| 1-(Perfluorobutyryl)imidazole; NSC 151966 | |
| ≥98.5% (GC); 98.5 to 101.0 wt. % (NT) |