missing translation for 'onlineSavingsMsg'
Learn More

N-Heptafluorobutyrylimidazole, ≥98.5% (GC); 98.5-101.0 wt. % (NT), MilliporeSigma™ Supelco™

Catalog No. 111019349
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
10 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019349 1 mL
111018038 10 x 1 mL
111018077 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. 111019349 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' 743821ML

Chemical Identifiers

MDL Number MFCD00014503
Synonym 1-(Perfluorobutyryl)imidazole; NSC 151966

Specifications

Density 1.490 g/mL (at 25°C)
CAS 32477-35-3
Boiling Point 161°C
Flash Point 77°C - Closed Cup
Linear Formula C7H3F7N2 O
Quantity 1 mL
Formula Weight 264.10
Percent Purity ≥98.5% (GC); 98.5 to 101.0 wt. % (NT)
Grade For GC derivatization