missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Formylglycine 98.0+%, TCI America™
$160.30 - $508.89
Chemical Identifiers
| CAS | 2491-15-8 |
|---|---|
| Molecular Formula | C3H5NO3 |
| Molecular Weight (g/mol) | 103.08 |
| MDL Number | MFCD00037360 |
| InChI Key | UGJBHEZMOKVTIM-UHFFFAOYSA-N |
| Synonym | n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine |
| PubChem CID | 75606 |
| IUPAC Name | 2-formamidoacetic acid |
| SMILES | OC(=O)CNC=O |
Chemical Identifiers
| 2491-15-8 | |
| 103.08 | |
| UGJBHEZMOKVTIM-UHFFFAOYSA-N | |
| 75606 | |
| OC(=O)CNC=O |
| C3H5NO3 | |
| MFCD00037360 | |
| n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine | |
| 2-formamidoacetic acid |
Specifications
| 2491-15-8 | |
| White | |
| MFCD00037360 | |
| n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine | |
| OC(=O)CNC=O | |
| 103.08 | |
| 103.08 | |
| Crystalline Powder |
| 147°C | |
| C3H5NO3 | |
| 5 g | |
| UGJBHEZMOKVTIM-UHFFFAOYSA-N | |
| 2-formamidoacetic acid | |
| 75606 | |
| ≥98.0% (T) | |
| N-Formylglycine |
Safety and Handling
RTECSNumber : MC0547000
TSCA : No