accessibility menu, dialog, popup

UserName

HOOHNO

CAS RN 2491-15-8

IUPAC Name: 2-formamidoacetic acid
Synonyms: glycine, N-formyl-
Molecular Weight (g/mol): 103.08
Molecular Formula: C3H5NO3
InChi Key: UGJBHEZMOKVTIM-UHFFFAOYSA-N
SMILES: OC(=O)CNC=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)