missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine 98.0+%, TCI America™

Supplier:  TCI America F012125G

 View more versions of this product

Catalog No. F012125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Formylglycine
147°C
C3H5NO3
25 g
UGJBHEZMOKVTIM-UHFFFAOYSA-N
2-formamidoacetic acid
75606
≥98.0% (T)
2491-15-8
White
MFCD00037360
n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine
OC(=O)CNC=O
103.08
103.08
Crystalline Powder

Chemical Identifiers

2491-15-8
103.08
UGJBHEZMOKVTIM-UHFFFAOYSA-N
75606
OC(=O)CNC=O
C3H5NO3
MFCD00037360
n-formylglycine, formylglycine, glycine, n-formyl, fgly, for-gly-oh, formylglycin, n-hydroxymethyleneglycine, formylamino acetic acid, n-formylglycin, n-formyl-glycine
2-formamidoacetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MC0547000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions