missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-methionine, 95%

CAS: 4289-98-9 | C6H11NO3S | 177.22 g/mol

$53.48 - $541.95

Chemical Identifiers

CAS 4289-98-9
Molecular Formula C6H11NO3S
Molecular Weight (g/mol) 177.22
MDL Number MFCD00021033
InChI Key PYUSHNKNPOHWEZ-YFKPBYRVSA-N
Synonym n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
PubChem CID 439750
ChEBI CHEBI:16552
IUPAC Name (2S)-2-formamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63493MD
View Documents
Thermo Scientific Chemicals
H63493MD
250 mg
Each for $53.48
Only null left
 
AAH6349303
View Documents
Thermo Scientific Chemicals
H6349303
1 g
Each for $128.51
Only null left
 
AAH6349306
View Documents
Thermo Scientific Chemicals
H6349306
5 g
Each for $541.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4289-98-9
177.22
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
439750
(2S)-2-formamido-4-methylsulfanylbutanoic acid
C6H11NO3S
MFCD00021033
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
CHEBI:16552
CSCC[C@H](NC=O)C(O)=O

Specifications

4289-98-9
MFCD00021033
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
(2S)-2-formamido-4-methylsulfanylbutanoic acid
177.22
CHEBI:16552
95%
N-Formyl-L-methionine
C6H11NO3S
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
CSCC[C@H](NC=O)C(O)=O
250 mg
439750
177.22
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only