accessibility menu, dialog, popup

UserName

H3CS(S)NHOOHO

CAS RN 4289-98-9

IUPAC Name: (2S)-2-formamido-4-(methylsulfanyl)butanoic acid
Synonyms: N-formylmethionine|fMet|For-Met-OH
Molecular Weight (g/mol): 177.22
Molecular Formula: C6H11NO3S
InChi Key: PYUSHNKNPOHWEZ-YFKPBYRVSA-N
SMILES: CSCC[C@H](NC=O)C(O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)