missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-methionine, 95%

CAS: 4289-98-9 | C6H11NO3S | 177.22 g/mol

Supplier:  Thermo Scientific Chemicals H6349306

 View more versions of this product

Catalog No. AAH6349306

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Formyl-L-methionine
C6H11NO3S
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
CSCC[C@H](NC=O)C(O)=O
5 g
439750
177.22
Solid
4289-98-9
MFCD00021033
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
(2S)-2-formamido-4-methylsulfanylbutanoic acid
177.22
CHEBI:16552
95%

Chemical Identifiers

4289-98-9
177.22
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
439750
(2S)-2-formamido-4-methylsulfanylbutanoic acid
C6H11NO3S
MFCD00021033
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
CHEBI:16552
CSCC[C@H](NC=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only