missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-O-trityl-D-serine, 95%
CAS: 212688-51-2 | C37H31NO5 | 569.657 g/mol
$284.22 - $1112.14
Chemical Identifiers
| CAS | 212688-51-2 |
|---|---|
| Molecular Formula | C37H31NO5 |
| Molecular Weight (g/mol) | 569.657 |
| MDL Number | MFCD00270546 |
| InChI Key | UCARTONYOJORBQ-UUWRZZSWSA-N |
| Synonym | fmoc-d-ser trt-oh, fmoc-o-trityl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethoxy propanoic acid, ambotzfaa1200, n-alpha-9-fluorenylmethyloxycarbonyl-o-trityl-d-serine |
| PubChem CID | 11621106 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)OCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6651506
|
Thermo Scientific Chemicals
H6651506 |
5 g |
Each for $284.22
|
|
|||||
|
AAH6651514
|
Thermo Scientific Chemicals
H6651514 |
25 g |
Each for $1,112.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 212688-51-2 | |
| 569.657 | |
| UCARTONYOJORBQ-UUWRZZSWSA-N | |
| 11621106 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)OCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| C37H31NO5 | |
| MFCD00270546 | |
| fmoc-d-ser trt-oh, fmoc-o-trityl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethoxy propanoic acid, ambotzfaa1200, n-alpha-9-fluorenylmethyloxycarbonyl-o-trityl-d-serine | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid |
Specifications
| 212688-51-2 | |
| MFCD00270546 | |
| UCARTONYOJORBQ-UUWRZZSWSA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid | |
| 569.657 | |
| 569.66 | |
| Solid |
| C37H31NO5 | |
| fmoc-d-ser trt-oh, fmoc-o-trityl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethoxy propanoic acid, ambotzfaa1200, n-alpha-9-fluorenylmethyloxycarbonyl-o-trityl-d-serine | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)OCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 | |
| 5 g | |
| 11621106 | |
| 95% | |
| N-Fmoc-O-trityl-D-serine |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only