missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-trityl-D-serine, 95%

CAS: 212688-51-2 | C37H31NO5 | 569.657 g/mol

Supplier:  Thermo Scientific Chemicals H6651506

 View more versions of this product

Catalog No. AAH6651506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-O-trityl-D-serine
C37H31NO5
fmoc-d-ser trt-oh, fmoc-o-trityl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethoxy propanoic acid, ambotzfaa1200, n-alpha-9-fluorenylmethyloxycarbonyl-o-trityl-d-serine
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)OCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
5 g
11621106
95%
212688-51-2
MFCD00270546
UCARTONYOJORBQ-UUWRZZSWSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid
569.657
569.66
Solid

Chemical Identifiers

212688-51-2
569.657
UCARTONYOJORBQ-UUWRZZSWSA-N
11621106
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)OCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
C37H31NO5
MFCD00270546
fmoc-d-ser trt-oh, fmoc-o-trityl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethoxy propanoic acid, ambotzfaa1200, n-alpha-9-fluorenylmethyloxycarbonyl-o-trityl-d-serine
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-trityloxypropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only