accessibility menu, dialog, popup

UserName

CAS RN 212688-51-2

OOOHHNOO

CAS RN 212688-51-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(triphenylmethoxy)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(triphenylmethoxy)propanoic acid
Molecular Weight (g/mol): 569.66
Molecular Formula: C37H31NO5
InChi Key: UCARTONYOJORBQ-UHFFFAOYNA-N
SMILES: OC(=O)C(COC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)