missing translation for 'onlineSavingsMsg'
Learn More

N-α-FMOC-N-delta-Trityl-L-glutamine, 95%, Thermo Scientific™

$48.45 - $149.05

Chemical Identifiers

CAS 132327-80-1 , 75-09-2
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
InChI Key WDGICUODAOGOMO-DHUJRADRSA-N
Synonym fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
PubChem CID 10919157
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC440640050
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440640250
View Documents Promotion Details
N/A 25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

132327-80-1 , 75-09-2
610.71
fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
C39H34N2O5
WDGICUODAOGOMO-DHUJRADRSA-N
10919157
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

132327-80-1 , 75-09-2
100.0
97.5% min. (HPLC)
− 14.20
WDGICUODAOGOMO-DHUJRADRSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
10919157
95%
94.0
Authentic
C39H34N2O5
fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
610.71
610.71
N-α-FMOC-N-δ-Trityl-L-glutamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only