missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%

Catalog No. AC440640050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440640050 5 g
AC440640250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440640050 Supplier Thermo Scientific Chemicals Supplier No. 440640050

CAS: 132327-80-1 | C39H34N2O5 | 610.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132327-80-1 , 75-09-2
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
InChI Key WDGICUODAOGOMO-DHUJRADRSA-N
Synonym fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
PubChem CID 10919157
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 131.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation − 14.20
Formula Weight 610.71
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-α-FMOC-N-δ-Trityl-L-glutamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only