missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-valinol, 98%, Thermo Scientific Chemicals

$99.74 - $99.74

Chemical Identifiers

CAS 160885-98-3
Molecular Formula C20H23NO3
Molecular Weight (g/mol) 325.41
MDL Number MFCD00235961
InChI Key MYMGENAMKAPEMT-LJQANCHMSA-N
Synonym fmoc-valinol, fmoc-l-valinol, n-fmoc-l-valinol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-methylbutan-2-yl carbamate, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, n-9-fluorenylmethoxycarbonyl-l-valinol, s-2-fmoc-amino-3-methyl-1-butanol, s-2-9h-fluorene-9-ylmethoxycarbonylamino-3-methyl-1-butanol, s-2-9-fluorenylmethyloxycarbonyl amino-3-methyl-1-butanol, n-alpha-9-fluorenylmethyloxycarbonyl-l-valinol
PubChem CID 688273
IUPAC Name (9H-fluoren-9-yl)methyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
SMILES CC(C)[C@@H](CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6632303
View Documents
Thermo Scientific Chemicals
H6632303
1 g
Each for $99.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

160885-98-3
325.41
MYMGENAMKAPEMT-LJQANCHMSA-N
688273
CC(C)[C@@H](CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C20H23NO3
MFCD00235961
fmoc-valinol, fmoc-l-valinol, n-fmoc-l-valinol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-methylbutan-2-yl carbamate, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, n-9-fluorenylmethoxycarbonyl-l-valinol, s-2-fmoc-amino-3-methyl-1-butanol, s-2-9h-fluorene-9-ylmethoxycarbonylamino-3-methyl-1-butanol, s-2-9-fluorenylmethyloxycarbonyl amino-3-methyl-1-butanol, n-alpha-9-fluorenylmethyloxycarbonyl-l-valinol
(9H-fluoren-9-yl)methyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate

Specifications

160885-98-3
MFCD00235961
fmoc-valinol, fmoc-l-valinol, n-fmoc-l-valinol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-methylbutan-2-yl carbamate, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate, n-9-fluorenylmethoxycarbonyl-l-valinol, s-2-fmoc-amino-3-methyl-1-butanol, s-2-9h-fluorene-9-ylmethoxycarbonylamino-3-methyl-1-butanol, s-2-9-fluorenylmethyloxycarbonyl amino-3-methyl-1-butanol, n-alpha-9-fluorenylmethyloxycarbonyl-l-valinol
CC(C)[C@@H](CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
325.41
325.41
Solid
C20H23NO3
1 g
MYMGENAMKAPEMT-LJQANCHMSA-N
(9H-fluoren-9-yl)methyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
688273
98%
N-Fmoc-L-valinol
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only