accessibility menu, dialog, popup

UserName

H3CCH3(S)OHNHOO

CAS RN 160885-98-3

IUPAC Name: (9H-fluoren-9-yl)methyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate|FMOC-L-VALINOL
Molecular Weight (g/mol): 325.41
Molecular Formula: C20H23NO3
InChi Key: MYMGENAMKAPEMT-LJQANCHMSA-N
SMILES: CC(C)[C@@H](CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results