missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-valine, 98%

Catalog No. AAB2103014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2103014 25 g
AAB2103006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2103014 Supplier Thermo Scientific Chemicals Supplier No. B2103014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68858-20-8 | C20H21NO4 | 339.39 g/mol

It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Fmoc-valine (in addition to the other amino acids) is commonly used to synthesize 4-thiazolidinones (e.g. (E)-5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one [E925745]) and 4-metathiazanones as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Fmoc-valine (in addition to the other amino acids) is commonly used to synthesize 4-thiazolidinones (e.g. (E)-5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one [E925745]) and 4-metathiazanones as well.

Solubility
Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 68858-20-8
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00037124
InChI Key UGNIYGNGCNXHTR-SFHVURJKSA-N
Synonym fmoc-l-valine, fmoc-val-oh, fmoc-l-val-oh, n-fmoc-l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-l-valine, n-9-fluorenylmethoxycarbonyl-l-valine, fmoc-valine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-alpha-fmoc-l-valine, s-n-9h-fluoren-9-ylmethoxy carbonyl valine
PubChem CID 688217
SMILES CC(C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 142°C to 147°C
Quantity 25 g
Beilstein 2177443
Optical Rotation −17° (c=1 in DMF)
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-valine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 272-515-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only