missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-proline, 98%

Catalog No. p-7025548
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2108106 5 g
AAB2108114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2108106 Supplier Thermo Scientific Chemicals Supplier No. B2108106

CAS: 71989-31-6 | C20H19NO4 | 337.38 g/mol

It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

Solubility
Soluble in methanol. (almost transparency.)

Notes
Store at 2-8°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 71989-31-6
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.38
MDL Number MFCD00037122
InChI Key ZPGDWQNBZYOZTI-UHFFFAOYNA-N
Synonym fmoc-pro-oh, fmoc-l-proline, fmoc-l-pro-oh, n-fmoc-l-proline, n-9-fluorenylmethoxycarbonyl-l-proline, s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-proline, fmoc-pro, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, 9-fluorenylmethoxycarbonyl-l-proline
PubChem CID 688135
IUPAC Name (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 113°C to 117°C
Quantity 5 g
Beilstein 3596735
Solubility Information Soluble in methanol. (almost transparency.)
Optical Rotation −40° (c=1 in Ethyl Acetate)
Formula Weight 337.38
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-proline
EINECSNumber 276-259-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only