missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-norleucine, 98%, Thermo Scientific Chemicals

Catalog No. AAB2247503
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2247503 1 g
AAB2247514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2247503 Supplier Thermo Scientific Chemicals Supplier No. B2247503

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77284-32-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037537
InChI Key VCFCFPNRQDANPN-IBGZPJMESA-N
Synonym fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
PubChem CID 7009636
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 140°C to 142°C
Quantity 1 g
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norleucine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only