missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-homophenylalanine, 95%, Thermo Scientific Chemicals
$79.27 - $458.97
Chemical Identifiers
| CAS | 132684-59-4 |
|---|---|
| Molecular Formula | C25H23NO4 |
| Molecular Weight (g/mol) | 401.46 |
| MDL Number | MFCD00077046 |
| InChI Key | CIHPCIUGLIZADU-QHCPKHFHSA-N |
| Synonym | fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid |
| PubChem CID | 2761467 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid |
| SMILES | OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH51973MD
|
Thermo Scientific Chemicals
H51973MD |
250 mg |
Each for $79.27
|
|
|||||
|
AAH5197306
|
Thermo Scientific Chemicals
H5197306 |
5 g |
Each for $458.97
|
|
|||||
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 132684-59-4 | |
| 401.46 | |
| CIHPCIUGLIZADU-QHCPKHFHSA-N | |
| 2761467 | |
| OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C25H23NO4 | |
| MFCD00077046 | |
| fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid |
Specifications
| 132684-59-4 | |
| MFCD00077046 | |
| 4847669 | |
| Slightly soluble in water. | |
| OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 401.46 | |
| 401.46 | |
| N-Fmoc-L-homophenylalanine |
| C25H23NO4 | |
| 250 mg | |
| fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid | |
| CIHPCIUGLIZADU-QHCPKHFHSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid | |
| 2761467 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only