missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-homophenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5197306
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| N-Fmoc-L-homophenylalanine | |
| C25H23NO4 | |
| 5 g | |
| fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid | |
| CIHPCIUGLIZADU-QHCPKHFHSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid | |
| 2761467 | |
| 95% |
| 132684-59-4 | |
| MFCD00077046 | |
| 4847669 | |
| Slightly soluble in water. | |
| OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 401.46 | |
| 401.46 |
Chemical Identifiers
| 132684-59-4 | |
| 401.46 | |
| CIHPCIUGLIZADU-QHCPKHFHSA-N | |
| 2761467 | |
| OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C25H23NO4 | |
| MFCD00077046 | |
| fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only