missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homovaline, 95%

CAS: 172695-33-9 | C21H23NO4 | 353.42 g/mol

$204.78 - $716.71

Chemical Identifiers

CAS 172695-33-9
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD01862853
InChI Key KFDMTAMWOIQTOB-LJQANCHMSA-N
Synonym fmoc-l-beta-homovaline, fmoc-, a-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, r-n-fmoc-3-amino-4-methylpentanoic acid, fmoc-beta-leu-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, a-leucine, fmoc-l-, a-leucine, fmoc-l-beta-leucine
PubChem CID 2761560
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
SMILES CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH52182MD
View Documents
Thermo Scientific Chemicals
H52182MD
250 mg
Each for $204.78
Only null left
 
AAH5218203
View Documents
Thermo Scientific Chemicals
H5218203
1 g
Each for $716.71
Only null left
 
Description

Description

N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

172695-33-9
353.42
KFDMTAMWOIQTOB-LJQANCHMSA-N
2761560
CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C21H23NO4
MFCD01862853
fmoc-l-beta-homovaline, fmoc-, a-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, r-n-fmoc-3-amino-4-methylpentanoic acid, fmoc-beta-leu-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, a-leucine, fmoc-l-, a-leucine, fmoc-l-beta-leucine
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid

Specifications

172695-33-9
MFCD01862853
7388393
Slightly soluble in water.
CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
353.42
353.42
N-Fmoc-L-beta-homovaline
C21H23NO4
250 mg
fmoc-l-beta-homovaline, fmoc-, a-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, r-n-fmoc-3-amino-4-methylpentanoic acid, fmoc-beta-leu-oh, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, a-leucine, fmoc-l-, a-leucine, fmoc-l-beta-leucine
KFDMTAMWOIQTOB-LJQANCHMSA-N
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
2761560
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only