Learn More
N-Fmoc-L-beta-homoproline, 95%
CAS: 193693-60-6 | C21H21NO4 | 351.402 g/mol
Supplier: Thermo Scientific Chemicals H5206003
Description
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| N-Fmoc-L-beta-homoproline | |
| C21H21NO4 | |
| 1 g | |
| fmoc-l-beta-homoproline, fmoc-beta-hopro-oh, s-2-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidin-2-yl acetic acid, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidin-2-yl acetic acid, 2-2s-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidin-2-yl acetic acid, fmoc-d-beta-homoproline, fmoc-hopro-oh, fmoc-beta-homopro-oh, fmoc-l-beta-hpro-oh, fmoc-i 2-hopro-oh | |
| C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)CC(=O)O | |
| 351.402 | |
| 351.4 |
| 193693-60-6 | |
| MFCD01863058 | |
| 7798553 | |
| ZNIGOUDZWCDFFC-AWEZNQCLSA-N | |
| 2-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidin-2-yl]acetic acid | |
| 2761542 | |
| 95% |
Chemical Identifiers
| 193693-60-6 | |
| 351.402 | |
| ZNIGOUDZWCDFFC-AWEZNQCLSA-N | |
| 2761542 | |
| C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)CC(=O)O |
| C21H21NO4 | |
| MFCD01863058 | |
| fmoc-l-beta-homoproline, fmoc-beta-hopro-oh, s-2-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidin-2-yl acetic acid, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidin-2-yl acetic acid, 2-2s-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidin-2-yl acetic acid, fmoc-d-beta-homoproline, fmoc-hopro-oh, fmoc-beta-homopro-oh, fmoc-l-beta-hpro-oh, fmoc-i 2-hopro-oh | |
| 2-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidin-2-yl]acetic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only