missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homoproline, 95%

Catalog No. p-7034377
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52060MD 250 mg
AAH5206003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52060MD Supplier Thermo Scientific Chemicals Supplier No. H52060MD
Only null left

CAS: 193693-60-6 | C21H21NO4 | 351.402 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 193693-60-6
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD01863058
InChI Key ZNIGOUDZWCDFFC-AWEZNQCLSA-N
Synonym fmoc-l-beta-homoproline, fmoc-beta-hopro-oh, s-2-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidin-2-yl acetic acid, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidin-2-yl acetic acid, 2-2s-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidin-2-yl acetic acid, fmoc-d-beta-homoproline, fmoc-hopro-oh, fmoc-beta-homopro-oh, fmoc-l-beta-hpro-oh, fmoc-i 2-hopro-oh
PubChem CID 2761542
IUPAC Name 2-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidin-2-yl]acetic acid
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)CC(=O)O

Specifications

Quantity 250 mg
Beilstein 7798553
Formula Weight 351.4
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homoproline
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only