missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%

Catalog No. p-7034502
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52188MD 250 mg
AAH5218803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52188MD Supplier Thermo Scientific Chemicals Supplier No. H52188MD

CAS: 203854-49-3 | C25H29NO6 | 439.508 g/mol

N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 203854-49-3
Molecular Formula C25H29NO6
Molecular Weight (g/mol) 439.508
MDL Number MFCD01863053
InChI Key XPCDWOCHPTYDPV-INIZCTEOSA-N
Synonym fmoc-beta-hoglu otbu-oh, fmoc-l-beta-homoglutamic acid 6-tert-butyl ester, fmoc-l-beta-homoglutamic acid otbu, fmoc-beta-homoglu otbu-oh, s-3-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxy-6-oxohexanoic acid, 3s-6-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-6-oxohexanoic acid, n-beta-9-fluorenylmethoxycarbonyl-l-beta-homoglutamic acid delta-t-butyl ester, ambotzfaa6670, fmoc-b-hoglu otbu-oh, fmoc-l-?-homoglutamic acid otbu
PubChem CID 2761518
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxy]-6-oxohexanoic acid
SMILES CC(C)(C)OC(=O)CCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 115°C to 120°C
Quantity 250 mg
Beilstein 8019724
Solubility Information Slightly soluble in water.
Formula Weight 439.51
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only