missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-alanine monohydrate, 98%

Catalog No. p-7025562
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2110706 5 g
AAB2110714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2110706 Supplier Thermo Scientific Chemicals Supplier No. B2110706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 207291-76-7 | C18H19NO5 | 329.35 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 207291-76-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
MDL Number MFCD00037139
InChI Key GAPWKFLOMOFHGO-UHFFFAOYNA-N
Synonym n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate
PubChem CID 18528118
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate
SMILES O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∼130°C
Quantity 5 g
Beilstein 2225975
Solubility Information Insoluble in water.
Optical Rotation −19° (c=1 in DMF)
Formula Weight 329.36 (311.34 Anhydrous)
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-alanine monohydrate
EINECSNumber 252-660-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only