accessibility menu, dialog, popup

UserName

CAS RN 207291-76-7

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid hydrate
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid hydrate
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: GAPWKFLOMOFHGO-UHFFFAOYNA-N
SMILES: O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results