missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-pyridyl)-L-alanine, 95%

CAS: 169555-95-7 | C23H20N2O4 | 388.423 g/mol

Supplier:  Thermo Scientific Chemicals H5217206

 View more versions of this product

Catalog No. AAH5217206


Only null left
Explore available promotions

Description

Description

Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications

Specifications

N-Fmoc-3-(4-pyridyl)-L-alanine
C23H20N2O4
5 g
fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine
SCSSXJVRZMQUKA-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
978322
95%
169555-95-7
MFCD00672566
7316905
Soluble in 0.1g in 4mL DMF(clearly soluble).
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O
388.423
388.42

Chemical Identifiers

169555-95-7
388.423
SCSSXJVRZMQUKA-NRFANRHFSA-N
978322
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O
C23H20N2O4
MFCD00672566
fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only