missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-pyridyl)-L-alanine, 95%

Catalog No. p-7034486
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52172MD 250 mg
AAH5217203 1 g
AAH5217206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52172MD Supplier Thermo Scientific Chemicals Supplier No. H52172MD
Only null left

CAS: 169555-95-7 | C23H20N2O4 | 388.423 g/mol

Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 169555-95-7
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00672566
InChI Key SCSSXJVRZMQUKA-NRFANRHFSA-N
Synonym fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine
PubChem CID 978322
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O

Specifications

Quantity 250 mg
Beilstein 7316905
Solubility Information Soluble in 0.1g in 4mL DMF(clearly soluble).
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-pyridyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only