missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(2-naphthyl)-L-alanine, 95%

CAS: 112883-43-9 | C28H22NO4 | 436.49 g/mol

Supplier:  Thermo Scientific Chemicals H5217906

 View more versions of this product

Catalog No. AAH5217906


Only null left
Explore available promotions

Description

Description

N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

N-Fmoc-3-(2-naphthyl)-L-alanine
C28H22NO4
5 g
fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347
JYUTZJVERLGMQZ-SANMLTNESA-M
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
2734452
95%
112883-43-9
MFCD00144886
7668973
Slightly soluble in water.
[O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
436.49
437.49

Chemical Identifiers

112883-43-9
436.49
JYUTZJVERLGMQZ-SANMLTNESA-M
2734452
[O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C28H22NO4
MFCD00144886
fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only