missing translation for 'onlineSavingsMsg'
Learn More

N-Fluorobenzenesulfonimide 97.0+%, TCI America™

Catalog No. p-7129619
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03355G 5 g
F033525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03355G Supplier TCI America Supplier No. F03355G
Only null left

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.333
MDL Number MFCD00144885
InChI Key RLKHFSNWQCZBDC-UHFFFAOYSA-N
Synonym n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
PubChem CID 588007
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 315.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Fluorobenzenesulfonimide [Fluorinating Reagent]