missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fluoro-N'-(chloromethyl)triethylenediamine Bis(tetrafluoroborate) 95.0+%, TCI America™
Supplier: TCI America F0358100G
Specifications
| N-Fluoro-N′-(chloromethyl)triethylenediamine Bis(tetrafluoroborate) | |
| 175°C | |
| C7H14B2ClF9N2 | |
| 100 g | |
| selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
| F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 | |
| 354.26 | |
| 354.26 | |
| Crystalline Powder |
| 140681-55-6 | |
| White-Yellow | |
| MFCD00142607 | |
| 3088 | |
| TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
| 4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide) | |
| 2724933 | |
| ≥95.0% (T) |
Chemical Identifiers
| 140681-55-6 | |
| 354.26 | |
| TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
| 2724933 | |
| F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 |
| C7H14B2ClF9N2 | |
| MFCD00142607 | |
| selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
| 4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide) |
Safety and Handling
TSCA : Yes