accessibility menu, dialog, popup

UserName

CAS RN 140681-55-6

IUPAC Name: 4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide)
Synonyms: 4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium ditetrafluoroborate|selectfluor
Molecular Weight (g/mol): 354.26
Molecular Formula: C7H14B2ClF9N2
InChi Key: TXRPHPUGYLSHCX-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)