missing translation for 'onlineSavingsMsg'
Learn More

N-Fluoro-N'-(chloromethyl)triethylenediamine Bis(tetrafluoroborate) 95.0+%, TCI America™

Catalog No. p-7129635
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03585G 5 g
F035825G 25 g
F0358100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F03585G Supplier TCI America Supplier No. F03585G
Only null left

Chemical Identifiers

CAS 140681-55-6
Molecular Formula C7H14B2ClF9N2
Molecular Weight (g/mol) 354.26
MDL Number MFCD00142607
InChI Key TXRPHPUGYLSHCX-UHFFFAOYSA-N
Synonym selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate
PubChem CID 2724933
IUPAC Name 4-(chloromethyl)-1-fluoro-1,4-diazabicyclo[2.2.2]octane-1,4-diium; bis(tetrafluoroboranuide)
SMILES F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 5 g
UN Number 3088
Formula Weight 354.26
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Fluoro-N′-(chloromethyl)triethylenediamine Bis(tetrafluoroborate)