missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethylpropylamine 97.0+%, TCI America™
Supplier: TCI America E0351250ML
Specifications
| N-Ethylpropylamine | |
| Yellow | |
| C5H13N | |
| 250 mL | |
| n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine | |
| CCCNCC | |
| 87.166 | |
| 87.17 | |
| Liquid |
| 20193-20-8 | |
| 79°C | |
| MFCD00015198 | |
| 2733 | |
| XCVNDBIXFPGMIW-UHFFFAOYSA-N | |
| N-ethylpropan-1-amine | |
| 88398 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 20193-20-8 | |
| 87.166 | |
| XCVNDBIXFPGMIW-UHFFFAOYSA-N | |
| 88398 | |
| CCCNCC |
| C5H13N | |
| MFCD00015198 | |
| n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine | |
| N-ethylpropan-1-amine |
Safety and Handling
TSCA : Yes