missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylpropylamine 97.0+%, TCI America™

Catalog No. p-7128954
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E035125ML 25 mL
E0351250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E035125ML Supplier TCI America Supplier No. E035125ML

Chemical Identifiers

CAS 20193-20-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015198
InChI Key XCVNDBIXFPGMIW-UHFFFAOYSA-N
Synonym n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
PubChem CID 88398
IUPAC Name N-ethylpropan-1-amine
SMILES CCCNCC

Specifications

Color Yellow
Boiling Point 79°C
Quantity 25 mL
UN Number 2733
Formula Weight 87.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethylpropylamine