accessibility menu, dialog, popup

UserName

H3CH2NCH3

CAS RN 20193-20-8

IUPAC Name: ethyl(propyl)azanium
Synonyms: ethyl(propyl)azanium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: XCVNDBIXFPGMIW-UHFFFAOYSA-O
SMILES: CCC[NH2+]CC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)