missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethylpropionamide 99.0+%, TCI America™
Supplier: TCI America E032825G
Specifications
| N-Ethylpropionamide | |
| Yellow | |
| C5H11NO | |
| 25 g | |
| ABMDIECEEGFXNC-UHFFFAOYSA-N | |
| N-ethylpropanamide | |
| 521324 | |
| ≥99.0% (GC) |
| 5129-72-6 | |
| 116°C | |
| MFCD00059388 | |
| N-Propionylethylamine | |
| CCNC(=O)CC | |
| 101.15 | |
| 101.15 | |
| Liquid |
Chemical Identifiers
| 5129-72-6 | |
| 101.15 | |
| ABMDIECEEGFXNC-UHFFFAOYSA-N | |
| 521324 | |
| CCNC(=O)CC |
| C5H11NO | |
| MFCD00059388 | |
| N-Propionylethylamine | |
| N-ethylpropanamide |
Safety and Handling
TSCA : No