missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylpropionamide 99.0+%, TCI America™

Catalog No. p-7128939
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03285G 5 g
E032825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03285G Supplier TCI America Supplier No. E03285G

Chemical Identifiers

CAS 5129-72-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00059388
InChI Key ABMDIECEEGFXNC-UHFFFAOYSA-N
Synonym N-Propionylethylamine
PubChem CID 521324
IUPAC Name N-ethylpropanamide
SMILES CCNC(=O)CC

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 101.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethylpropionamide