accessibility menu, dialog, popup

UserName

CAS RN 5129-72-6

IUPAC Name: N-ethylpropanamide
Synonyms: N-ethylpropanamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: ABMDIECEEGFXNC-UHFFFAOYSA-N
SMILES: CCNC(=O)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results