missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylbenzylamine, 95%

CAS: 14321-27-8 | C9H13N | 135.21 g/mol

Supplier:  Thermo Scientific Chemicals L05651AP

 View more versions of this product

Catalog No. AAL05651AP


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Ethylbenzylamine
0.91
66°C (150°F)
1.511
500 mL
386023
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
CCNCC1=CC=CC=C1
135.21
135.21
14321-27-8
191°C to 194°C
C9H13N
MFCD00009031
UN2735
Air Sensitive
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
N-benzylethanamine
84352
97%

Chemical Identifiers

14321-27-8
135.21
HVAAHUDGWQAAOJ-UHFFFAOYSA-N
84352
CCNCC1=CC=CC=C1
C9H13N
MFCD00009031
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
N-benzylethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 238-265-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only