missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylbenzylamine, 95%

Catalog No. AAL05651AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL05651AC 25 mL
AAL05651AE 100 mL
AAL05651AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL05651AC Supplier Thermo Scientific Chemicals Supplier No. L05651AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14321-27-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14321-27-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00009031
InChI Key HVAAHUDGWQAAOJ-UHFFFAOYSA-N
Synonym n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
PubChem CID 84352
IUPAC Name N-benzylethanamine
SMILES CCNCC1=CC=CC=C1

Specifications

Density 0.91
Boiling Point 191°C to 194°C
Flash Point 66°C (150°F)
Refractive Index 1.511
Quantity 25 mL
UN Number UN2735
Beilstein 386023
Sensitivity Air Sensitive
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material N-Ethylbenzylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 238-265-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only