missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-o-toluidine 98.0+%, TCI America™
$36.57 - $238.72
Chemical Identifiers
| CAS | 94-68-8 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00026695 |
| InChI Key | MWOUGPLLVVEUMM-UHFFFAOYSA-N |
| Synonym | n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine |
| PubChem CID | 7201 |
| IUPAC Name | N-ethyl-2-methylaniline |
| SMILES | CCNC1=CC=CC=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E017725ML
|
TCI America
E017725ML |
25 mL |
Each for $36.57
|
|
|||||
|
E0177500ML
|
TCI America
E0177500ML |
500 mL |
Each for $238.72
|
|
|||||
Chemical Identifiers
| 94-68-8 | |
| 135.21 | |
| MWOUGPLLVVEUMM-UHFFFAOYSA-N | |
| 7201 | |
| CCNC1=CC=CC=C1C |
| C9H13N | |
| MFCD00026695 | |
| n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine | |
| N-ethyl-2-methylaniline |
Specifications
| 94-68-8 | |
| 214°C | |
| MFCD00026695 | |
| 2754 | |
| MWOUGPLLVVEUMM-UHFFFAOYSA-N | |
| N-ethyl-2-methylaniline | |
| 7201 | |
| ≥98.0% (GC,T) | |
| N-Ethyl-o-toluidine |
| Yellow | |
| C9H13N | |
| 25 mL | |
| n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine | |
| CCNC1=CC=CC=C1C | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0187&(3)-0193
TSCA : Yes