missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-o-toluidine 98.0+%, TCI America™
Supplier: TCI America E0177500ML
Specifications
| N-Ethyl-o-toluidine | |
| Yellow | |
| C9H13N | |
| 500 mL | |
| n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine | |
| CCNC1=CC=CC=C1C | |
| 135.21 | |
| 135.21 | |
| Liquid |
| 94-68-8 | |
| 214°C | |
| MFCD00026695 | |
| 2754 | |
| MWOUGPLLVVEUMM-UHFFFAOYSA-N | |
| N-ethyl-2-methylaniline | |
| 7201 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 94-68-8 | |
| 135.21 | |
| MWOUGPLLVVEUMM-UHFFFAOYSA-N | |
| 7201 | |
| CCNC1=CC=CC=C1C |
| C9H13N | |
| MFCD00026695 | |
| n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine | |
| N-ethyl-2-methylaniline |
Safety and Handling
EINECSNumber : (3)-0187&(3)-0193
TSCA : Yes