missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-D-glucamine 98.0+%, TCI America™
$33.56 - $136.08
Chemical Identifiers
| CAS | 14216-22-9 |
|---|---|
| Molecular Formula | C8H19NO5 |
| Molecular Weight (g/mol) | 209.24 |
| MDL Number | MFCD03789564 |
| InChI Key | IKXCHOUDIPZROZ-LXGUWJNJSA-N |
| PubChem CID | 84266 |
| IUPAC Name | (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol |
| SMILES | CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO |
Chemical Identifiers
| 14216-22-9 | |
| 209.24 | |
| IKXCHOUDIPZROZ-LXGUWJNJSA-N | |
| (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol |
| C8H19NO5 | |
| MFCD03789564 | |
| 84266 | |
| CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO |
Specifications
| 14216-22-9 | |
| White | |
| MFCD03789564 | |
| IKXCHOUDIPZROZ-LXGUWJNJSA-N | |
| (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol | |
| 84266 | |
| ≥98.0% (T) | |
| N-Ethyl-D-glucamine |
| 139°C | |
| C8H19NO5 | |
| 25 g | |
| CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO | |
| 209.24 | |
| 209.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0209
TSCA : No