accessibility menu, dialog, popup

UserName

CAS RN 14216-22-9

H3CNH(S)OH(R)OH(R)OH(R)OHOH

CAS RN 14216-22-9

IUPAC Name: (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol
Synonyms: (2R,3R,4R,5S)-6-(ethylamino)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol): 209.24
Molecular Formula: C8H19NO5
InChi Key: IKXCHOUDIPZROZ-LXGUWJNJSA-N
SMILES: CCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results