missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonyl-5-nitro-o-toluidine 96.0+%, TCI America™

Catalog No. E03495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03495G 5 g
E034925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03495G Supplier TCI America Supplier No. E03495G
Only null left

Chemical Identifiers

CAS 16648-52-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00059827
InChI Key AYJFWTBDUMVOBQ-UHFFFAOYSA-N
Synonym N-Carbethoxy-5-nitro-o-toluidine, Ethyl 2-Methyl-5-nitrophenylcarbamate, N-(5-Nitro-2-methylphenyl)urethan
PubChem CID 601143
IUPAC Name ethyl N-(2-methyl-5-nitrophenyl)carbamate
SMILES CCOC(=O)NC1=C(C)C=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 135°C
Quantity 5 g
Formula Weight 224.22
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonyl-5-nitro-o-toluidine