accessibility menu, dialog, popup

UserName

H3COONHNOOH3C

CAS RN 16648-52-5

IUPAC Name: ethyl N-(2-methyl-5-nitrophenyl)carbamate
Synonyms: ethyl N-(2-methyl-5-nitrophenyl)carbamate
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: AYJFWTBDUMVOBQ-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1=C(C)C=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results