missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-epsilon-Acetyl-L-lysine, 99%, MP Biomedicals™
$155.14
Chemical Identifiers
| CAS | 692-04-6 |
|---|---|
| Molecular Formula | C8H16N2O3 |
| Molecular Weight (g/mol) | 188.23 |
| MDL Number | MFCD00002639 |
| InChI Key | DTERQYGMUDWYAZ-ZETCQYMHSA-N |
| Synonym | n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine |
| PubChem CID | 92832 |
| ChEBI | CHEBI:17752 |
| SMILES | CC(=O)NCCCC[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15023501
|
N/A | 1 g |
Each for $155.14
|
|
|||||
Chemical Identifiers
| C8H16N2O3 | |
| MFCD00002639 | |
| n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine | |
| CHEBI:17752 |
Specifications
| 692-04-6 | |
| 99% | |
| MFCD00002639 | |
| DTERQYGMUDWYAZ-ZETCQYMHSA-N | |
| (2S)-2-amino-6-acetamidohexanoic acid | |
| 92832 | |
| 188.3 | |
| N-epsilon-acetyl-L-lysine |
| White | |
| C8H16N2O3 | |
| n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine | |
| CC(=O)NCCCC[C@H](N)C(O)=O | |
| 188.23 | |
| CHEBI:17752 | |
| Powder |
Safety and Handling
Recommended Storage : Store at 0°C.