accessibility menu, dialog, popup

UserName

H3COHN(S)NH2OHO

CAS RN 692-04-6

IUPAC Name: (2S)-2-amino-6-acetamidohexanoic acid
Synonyms: N6-acetyllysine|ω-N-acetyl-L-lysine|epsilon-N-acetyllysine|N-epsilon-acetyllysine|ne-acetyllysine|epsilon-acetyl-L-lysine|N-epsilon-acetyl-L-lysine|ne-acetyl-L-lysine|N(epsilon)-acetyl-L-lysine|epsilon-N-acetyl-L-lysine
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: DTERQYGMUDWYAZ-ZETCQYMHSA-N
SMILES: CC(=O)NCCCC[C@H](N)C(O)=O
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)