missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl methyl sulfide, Thermo Scientific™

Catalog No. p-7049579
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1487706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1487706 Supplier Thermo Scientific Chemicals Supplier No. 1487706

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 3698-89-3
Molecular Formula C13H28S
Molecular Weight (g/mol) 216.43
MDL Number MFCD00015073
InChI Key KJWHJDGMOQJLGF-UHFFFAOYSA-N
Synonym dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u
PubChem CID 77287
IUPAC Name 1-methylsulfanyldodecane
SMILES CCCCCCCCCCCCSC

Specifications

Boiling Point 163°C to 165°C (19 mmHg)
Odor Sulphurous
Refractive Index 1.4608
Linear Formula CH3(CH2)11SCH3
Quantity 5 g
Formula Weight 216.41
Physical Form Liquid
Chemical Name or Material n-Dodecyl methyl sulfide
EINECSNumber 223-028-7
TSCA Yes
Recommended Storage Ambient temperatures